Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173430.2 | NP_775606.1 | 3 | 333 | ||
GenBank mRNA | AJ511806.1 | - | 3 | 316 | ||
AK080624.1 | - | 1 | 143 | |||
AK135089.1 | - | 3 | 334 | |||
AK156156.1 | - | 3 | 331 | |||
AK166068.1 | - | 3 | 335 | |||
AK168308.1 | - | 3 | 331 | |||
BC029216.1 | - | 3 | 328 | |||
BC053072.1 | - | 3 | 316 | 5686286 | ||
BY014040.1 | - | 3 | 334 |