Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nlrp4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172896.2 | NP_766484.1 | 4, 5 | 2168 | ||
GenBank mRNA | AK054378.1 | - | 4, 5 | 2059 | ||
AK087843.1 | - | 4, 5 | 2169 | |||
AK135745.1 | - | 1, 2 | 982 | |||
AK136205.1 | - | 5, 6 | 2144 | |||
AY355341.1 | - | 4, 5 | 2169 | |||
AY596200.1 | - | 2, 3 | 1865 | |||
BC141342.1 | - | 4, 5 | 2068 | 9055954 | ||
BC141343.1 | - | 4, 5 | 2068 | 9055955 |