Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Awat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081136.1 | NP_001074605.1 | 1, 2 | 154 | ||
GenBank mRNA | AK137763.1 | - | 1, 2 | 151 | ||
BC147680.1 | - | 1, 2 | 125 | 9056653 | ||
BC147683.1 | - | 1, 2 | 125 | 9056656 | ||
BC147829.1 | - | 1, 2 | 83 | 9087546 | ||
BC147832.1 | - | 1, 2 | 83 | 9087549 |