Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Awat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081136.1 | NP_001074605.1 | 2 | 219 | ||
GenBank mRNA | AK137763.1 | - | 2 | 216 | ||
BC147680.1 | - | 2 | 190 | 9056653 | ||
BC147683.1 | - | 2 | 190 | 9056656 | ||
BC147829.1 | - | 2 | 148 | 9087546 | ||
BC147832.1 | - | 2 | 148 | 9087549 |