Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chodl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139134.3 | NP_624360.2 | 4, 5 | 1049 | ||
GenBank mRNA | AF311699.1 | - | 4, 5 | 872 | ||
AK014255.1 | - | 4, 5 | 760 | |||
AK031063.1 | - | 4, 5 | 1050 | |||
AK076523.1 | - | 4, 5 | 1049 | |||
AK140910.1 | - | 4, 5 | 1050 | |||
BC117071.1 | - | 4, 5 | 720 | 40126390 | ||
BC117073.1 | - | 4, 5 | 720 | 40126392 | ||
BC144738.1 | - | 4, 5 | 705 | 9053286 |