Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147776.4 | NP_680085.3 | 2 | 648 | ||
GenBank mRNA | AK030019.1 | - | 2 | 602 | ||
AK077240.1 | - | 2 | 619 | |||
AY030094.1 | - | 2 | 618 | |||
BC026919.1 | - | 2 | 607 | 3982749 | ||
BC036166.1 | - | 2 | 613 | 4985003 |