Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vwa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147776.4 | NP_680085.3 | 2 | 308 | ||
GenBank mRNA | AK030019.1 | - | 2 | 262 | ||
AK077240.1 | - | 2 | 279 | |||
AY030094.1 | - | 2 | 278 | |||
BC026919.1 | - | 2 | 267 | 3982749 | ||
BC036166.1 | - | 2 | 273 | 4985003 | ||
CX210560.1 | - | 2 | 308 |