Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144559.2 | NP_653142.2 | 4 | 581 | ||
GenBank mRNA | AF499613.1 | - | 4 | 549 | ||
AK076532.1 | - | 4 | 587 | |||
AK170920.1 | - | 4 | 553 | |||
BC027310.1 | - | 4 | 568 | 3674499 | ||
EU050648.1 | - | 4 | 527 |