Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278787.1 | NP_001265716.1 | 2 | 424 | ||
NM_001278788.1 | NP_001265717.1 | 2 | 417 | |||
NM_146262.3 | NP_666374.2 | 3 | 633 | |||
GenBank mRNA | CB173233.1 | - | 2 | 424 |