Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr846 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146282.2 | NP_666394.1 | 1 | 99 | ||
GenBank mRNA | BC134375.1 | - | 1 | 100 | 40135046 |