Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr732 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146665.2 | NP_666876.1 | 1 | 102 | ||
GenBank mRNA | BC120848.1 | - | 1 | 102 | 40129771 |