Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1298 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146886.1 | NP_667097.1 | 1 | 675 | ||
GenBank mRNA | BC104070.1 | - | 1 | 675 | ||
BC104071.1 | - | 1 | 675 | |||
BC119407.1 | - | 1 | 675 | |||
BC119408.1 | - | 1 | 674 | |||
BC126868.1 | - | 1 | 675 | 40044133 | ||
BC127217.1 | - | 1 | 675 | 40042024 | ||
BC127218.1 | - | 1 | 675 | 40042025 |