Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1242 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146968.2 | NP_667179.1 | 1 | 235 | ||
GenBank mRNA | BC120865.1 | - | 1 | 235 | 40129788 | |
BC125386.1 | - | 1 | 235 | 40129901 |