Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr1037 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011532.2 | NP_001011532.1 | 1 | 511 | ||
GenBank mRNA | BC147135.1 | - | 1 | 511 | 9056192 | |
BC147136.1 | - | 1 | 511 | 9056181 |