Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139051.3 | NP_620639.1 | 4 | 1050 | ||
GenBank mRNA | AB000490.1 | - | 4 | 1094 | ||
AF511594.1 | - | 4 | 880 | |||
AK134378.1 | - | 4 | 1051 | |||
AK144376.1 | - | 4 | 1011 | |||
AK156911.1 | - | 4 | 1018 | |||
BC110476.1 | - | 4 | 993 | 40047261 | ||
BC110477.1 | - | 4 | 993 | 40047262 | ||
D10584.1 | - | 3 | 1280 |