Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139051.3 | NP_620639.1 | 7 | 1452 | ||
GenBank mRNA | AB000490.1 | - | 7 | 1496 | ||
AF511594.1 | - | 7 | 1282 | |||
AK134378.1 | - | 7 | 1453 | |||
AK144376.1 | - | 7 | 1413 | |||
AK156911.1 | - | 7 | 1420 | |||
BC110476.1 | - | 7 | 1395 | 40047261 | ||
BC110477.1 | - | 7 | 1395 | 40047262 |