Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc27a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011989.4 | NP_036119.1 | 7 | 1244 | ||
GenBank mRNA | AF072759.1 | - | 5 | 546 | ||
AJ251113.1 | - | 6 | 957 | |||
AK009292.1 | - | 1 | 31 | |||
AK033519.1 | - | 3 | 188 | |||
AK036919.1 | - | 7 | 1246 | |||
AK155313.1 | - | 7 | 1213 | |||
AK155388.1 | - | 7 | 1214 | |||
AK170377.1 | - | 7 | 1248 | |||
AK213719.1 | - | 1 | 27 | |||
BC023114.1 | - | 7 | 1095 | 4158956 | ||
CT010312.1 | - | 6 | 957 |