Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyb5r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024195.2 | NP_077157.2 | 15, 16 | 1686 | ||
GenBank mRNA | AF338818.1 | - | 14, 15 | 1434 | ||
AK009272.1 | - | 1, 2 | 21 | |||
AK012855.1 | - | 14, 15 | 1324 | |||
AK078682.1 | - | 8, 9 | 892 | |||
AK090159.1 | - | 15, 16 | 1688 | |||
AK157312.1 | - | 14, 15 | 1690 | |||
AK167436.1 | - | 14, 15 | 1384 | |||
AK169937.1 | - | 14, 15 | 1684 | |||
AK169995.1 | - | 14, 15 | 1681 | |||
AK172252.1 | - | 14, 15 | 1639 | |||
AK207052.1 | - | 3, 4 | 341 | |||
AK219801.1 | - | 3, 4 | 321 | |||
AY321368.1 | - | 13, 14 | 1458 | |||
AY321369.1 | - | 12, 13 | 1305 | |||
BC002170.1 | - | 8, 9 | 886 | |||
BC025438.1 | - | 15, 16 | 1697 | 5038563 |