Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100z | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081159.1 | NP_001074628.1 | 1, 2 | 133 | ||
GenBank mRNA | BC147382.1 | - | 1, 2 | 199 | 9056440 | |
BC147383.1 | - | 1, 2 | 199 | 9056460 |