Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Robo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175549.4 | NP_780758.3 | 4, 5 | 655 | ||
GenBank mRNA | AK035110.1 | - | 4, 5 | 1529 | ||
AK085461.1 | - | 4, 5 | 1280 | |||
BC055333.1 | - | 4, 5 | 963 | 6409283 | ||
DQ533875.1 | - | 5, 6 | 935 | |||
DQ533876.1 | - | 4, 5 | 655 |