Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173741.3 | NP_776102.1 | 1 | 719 | ||
GenBank mRNA | AK078694.1 | - | 1 | 699 | ||
AK170253.1 | - | 1 | 699 | |||
BC037651.1 | - | 1 | 641 | 5148210 |