Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops7b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172974.2 | NP_766562.1 | 5 | 643 | ||
GenBank mRNA | AF071317.1 | - | 5 | 621 | ||
AK053615.1 | - | 4 | 513 | |||
AK081309.1 | - | 5 | 643 | |||
AK168740.1 | - | 5 | 602 | |||
AK170140.1 | - | 5 | 602 | |||
AK182444.1 | - | 2 | 265 | |||
BC012659.1 | - | 5 | 607 | 4017012 |