Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vsnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012038.4 | NP_036168.1 | 4 | 838 | ||
GenBank mRNA | AJ489331.1 | - | 4 | 802 | ||
AK005091.2 | - | 3 | 400 | |||
AK144454.1 | - | 4 | 768 | |||
AK160224.1 | - | 4 | 770 | |||
AY101375.1 | - | 3 | 540 | |||
BC046226.1 | - | 4 | 743 | 5692284 | ||
D21165.1 | - | 3 | 540 | |||
EU373446.1 | - | 3 | 540 |