Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012053.2 | NP_036183.1 | 4, 5 | 541 | ||
GenBank mRNA | AK131970.1 | - | 4, 5 | 533 | ||
AK145498.1 | - | 4, 5 | 528 | |||
AK146330.1 | - | 4, 5 | 528 | |||
AK151408.1 | - | 4, 5 | 528 | |||
AK152063.1 | - | 4, 5 | 535 | |||
AK167775.1 | - | 4, 5 | 528 | |||
AK168769.1 | - | 4, 5 | 526 | |||
BC043017.1 | - | 3, 4 | 509 | 5684024 | ||
U67771.1 | - | 3, 4 | 513 |