Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galntl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175032.3 | NP_778197.2 | 4 | 667 | ||
GenBank mRNA | AK080614.1 | - | 4 | 667 | ||
BC111102.1 | - | 5 | 710 | |||
GU220061.1 | - | 4 | 514 |