Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map1s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173013.3 | NP_766601.2 | 5 | 1280 | ||
GenBank mRNA | AK032300.1 | - | 5 | 1268 | ||
AK042159.1 | - | 5 | 1277 | |||
AK082854.1 | - | 5 | 1275 | |||
AK158860.1 | - | 3 | 1049 | |||
BC052828.1 | - | 1 | 127 | |||
BC058166.1 | - | 5 | 1261 | |||
BC079598.1 | - | 3 | 2641 | |||
BC150729.1 | - | 5 | 1322 | 9087640 | ||
DQ387862.1 | - | 5 | 1268 |