Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm4216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167166.1 | NP_001160638.1 | 1 | 704 |
Gene Symbol: Gm4133 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167158.1 | NP_001160630.1 | 1 | 710 |
Gene Symbol: Gm4177 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167167.1 | NP_001160639.1 | 1 | 704 |
Gene Symbol: Gm4187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167159.1 | NP_001160631.1 | 1 | 710 |
Gene Symbol: Vmn1r115 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166745.1 | NP_001160217.1 | 1 | 704 |
Gene Symbol: Vmn1r119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166708.1 | NP_001160180.1 | 1 | 710 |
Gene Symbol: Vmn1r127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166726.1 | NP_001160198.1 | 1 | 704 |