Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163311.1 | NP_001156783.1 | 3 | 458 | ||
NM_013761.4 | NP_038789.1 | 4 | 684 | |||
GenBank mRNA | AF148321.1 | - | 3 | 339 | ||
AK017235.1 | - | 4 | 564 | |||
AK028034.1 | - | 3 | 341 | |||
AK031687.1 | - | 4 | 556 | |||
AK043738.1 | - | 4 | 590 | |||
AK043819.1 | - | 3 | 364 | |||
AK080830.1 | - | 4 | 564 | |||
AK157122.1 | - | 3 | 609 | |||
AK170096.1 | - | 4 | 570 | |||
AK170471.1 | - | 4 | 570 | |||
BC011164.1 | - | 4 | 561 | 4195695 |