Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289898.1 | NP_001276827.1 | 6 | 738 | ||
NM_001289899.1 | NP_001276828.1 | 6 | 1210 | |||
NM_013767.6 | NP_038795.3 | 6 | 742 | |||
GenBank mRNA | AB028241.1 | - | 6 | 688 | ||
AB028736.2 | - | 6 | 688 | |||
AK158709.1 | - | 6 | 1211 | |||
AK201751.1 | - | 2 | 159 | |||
BC026127.1 | - | 6 | 691 | 4010696 |