Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Naglu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013792.2 | NP_038820.2 | 6 | 1181 | ||
GenBank mRNA | AK143546.1 | - | 6 | 1183 | ||
AK158774.1 | - | 4 | 1805 | |||
AK164125.1 | - | 6 | 1160 | |||
AK170758.1 | - | 6 | 1164 | |||
AK182283.1 | - | 1 | 141 | |||
AK185044.1 | - | 1 | 133 | |||
AK188704.1 | - | 2 | 173 | |||
AK189169.1 | - | 2 | 173 | |||
AK208303.1 | - | 2 | 173 | |||
BC055733.1 | - | 6 | 1155 | 6411530 | ||
U85247.1 | - | 6 | 1150 |