Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nelfe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045863.1 | NP_001039328.1 | 10 | 1157 | ||
NM_001045864.1 | NP_001039329.1 | 10 | 1243 | |||
NM_138580.2 | NP_613046.2 | 10 | 1031 | |||
GenBank mRNA | AI325150.1 | - | 2 | 375 | ||
AK011663.1 | - | 10 | 1016 | |||
AK012389.1 | - | 10 | 1048 | |||
AK131947.1 | - | 10 | 1015 | |||
AK147011.1 | - | 10 | 1139 | |||
BC034867.1 | - | 10 | 983 | 1495547 | ||
DV662511.1 | - | 7 | 776 |