Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Yipf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145353.2 | NP_663328.2 | 2 | 349 | ||
GenBank mRNA | AF197931.1 | - | 1 | 109 | ||
AK011822.1 | - | 2 | 336 | |||
AK042644.1 | - | 1 | 21 | |||
AK149957.1 | - | 2 | 182 | |||
BC011161.1 | - | 2 | 310 | |||
BC019384.1 | - | 2 | 330 | 5123736 |