Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nop16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178605.4 | NP_848720.1 | 5 | 557 | ||
GenBank mRNA | AK002696.1 | - | 5 | 505 | ||
AK003769.1 | - | 5 | 540 | |||
AK013251.1 | - | 5 | 558 | |||
AK088248.1 | - | 5 | 558 |