Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slco1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013797.5 | NP_038825.1 | 14 | 2178 | ||
GenBank mRNA | AF148218.1 | - | 14 | 2002 | ||
AF223067.1 | - | 14 | 2012 | |||
AK041125.1 | - | 15 | 2116 | |||
AY195868.1 | - | 14 | 2011 | |||
AY195869.1 | - | 14 | 2011 | |||
AY243579.1 | - | 14 | 2011 | |||
BC041147.1 | - | 14 | 1961 | 4975986 |