Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cabp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013877.3 | NP_038905.1 | 3 | 324 | ||
GenBank mRNA | AB079605.1 | - | 2 | 140 | ||
AF169161.1 | - | 3 | 285 | |||
AK044544.1 | - | 3 | 325 | |||
AK087416.1 | - | 3 | 185 | |||
BC018253.1 | - | 3 | 347 | 4504283 |