Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ulk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013881.4 | NP_038909.3 | 24, 25 | 3270 | ||
GenBank mRNA | AB019577.1 | - | 24, 25 | 2795 | ||
AF145922.1 | - | 24, 25 | 3163 | |||
AK122331.2 | - | 24, 25 | 3252 | |||
AK135607.1 | - | 1, 2 | 1334 | |||
AK146620.1 | - | 24, 25 | 3232 | |||
AK146893.1 | - | 24, 25 | 3271 | |||
AK148142.1 | - | 24, 25 | 3271 | |||
BC013691.1 | - | 22, 23 | 2520 | |||
BC046778.1 | - | 24, 25 | 3228 | 5709559 | ||
BC053029.1 | - | 24, 25 | 3228 | 5709559 |