Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ulk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013881.4 | NP_038909.3 | 19, 20 | 2506 | ||
GenBank mRNA | AB019577.1 | - | 19, 20 | 2031 | ||
AF145922.1 | - | 19, 20 | 2399 | |||
AK122331.2 | - | 19, 20 | 2488 | |||
AK146620.1 | - | 19, 20 | 2468 | |||
AK146893.1 | - | 19, 20 | 2507 | |||
AK148142.1 | - | 19, 20 | 2507 | |||
BC013691.1 | - | 17, 18 | 1756 | |||
BC046778.1 | - | 19, 20 | 2464 | 5709559 | ||
BC053029.1 | - | 19, 20 | 2464 | 5709559 |