Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxw2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164768.1 | NP_001158240.1 | 5 | 1163 | ||
NM_001164769.1 | NP_001158241.1 | 7 | 1222 | |||
NM_001164770.1 | NP_001158242.1 | 7 | 1130 | |||
NM_013890.4 | NP_038918.3 | 7 | 1222 | |||
GenBank mRNA | AF140683.1 | - | 5 | 1167 | ||
AK033129.1 | - | 7 | 1176 | |||
AK040764.1 | - | 7 | 1189 | |||
AK078815.1 | - | 7 | 1130 | |||
CB203503.1 | - | 2 | 158 | |||
X54352.1 | - | 6 | 1152 |