Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175668.4 | NP_783599.1 | 2 | 1361 | ||
GenBank mRNA | AK085829.1 | - | 2 | 1363 | ||
AY255582.1 | - | 1 | 61 | |||
BC099398.1 | - | 1 | 1236 | 30713585 |