Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pgbd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012311.2 | NP_001012311.2 | 4 | 1221 | ||
NM_001162919.1 | NP_001156391.1 | 5 | 1390 | |||
NM_001162920.1 | NP_001156392.1 | 5 | 1332 | |||
GenBank mRNA | AK076559.1 | - | 5 | 1283 | ||
BC059882.1 | - | 5 | 1323 | |||
BC089360.1 | - | 4 | 1069 | 30613267 |