Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E130114P18Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK053617.1 | - | 271 | |||
AK082601.1 | - | 2 | 113 | |||
AK086199.1 | - | 447 | ||||
AK086602.1 | - | 2 | 157 | |||
BC048947.1 | - | 3 | 230 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177023.1 | - | 447 | Chr. - - [-] on Build |