Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oacyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177028.3 | NP_796002.1 | 2 | 199 | ||
GenBank mRNA | AK030604.1 | - | 2 | 199 | ||
BC141265.1 | - | 2 | 196 | 9055877 |