Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177074.2 | NP_796048.2 | 1, 2 | 150 | ||
GenBank mRNA | AK034126.1 | - | 1, 2 | 159 | ||
AK041015.1 | - | 1, 2 | 48 | |||
AK043610.1 | - | 1, 2 | 159 | |||
AK139966.1 | - | 1, 2 | 100 | |||
CJ055756.1 | - | 1, 2 | 151 |