Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B430306N03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177083.4 | NP_796057.1 | 4 | 812 | ||
GenBank mRNA | AK036709.1 | - | 4 | 813 | ||
AK046672.1 | - | 813 | ||||
AK133242.1 | - | 4 | 1673 | |||
AK155881.1 | - | 4 | 828 | |||
AK170621.1 | - | 4 | 834 | |||
AK171230.1 | - | 4 | 2491 | |||
AK171587.1 | - | 4 | 813 | |||
AK171603.1 | - | 4 | 811 | |||
AK171605.1 | - | 4 | 828 | |||
AY522648.1 | - | 4 | 813 | |||
DQ087179.1 | - | 4 | 565 | |||
DQ087180.1 | - | 5 | 656 |