Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Negr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039094.3 | NP_001034183.1 | 6 | 1255 | ||
NM_177274.4 | NP_796248.1 | 6 | 1255 | |||
GenBank mRNA | AJ487032.1 | - | 6 | 875 | ||
AK035218.1 | - | 6 | 1091 | |||
AK122576.2 | - | 5 | 598 | |||
AK142298.1 | - | 6 | 949 | |||
AK147253.1 | - | 6 | 1256 | |||
BC146005.1 | - | 6 | 1040 | 40131308 | ||
BC146007.1 | - | 6 | 1039 | 40131310 |