Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Negr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039094.3 | NP_001034183.1 | 2 | 718 | ||
NM_177274.4 | NP_796248.1 | 2 | 718 | |||
GenBank mRNA | AJ487032.1 | - | 2 | 338 | ||
AK030503.1 | - | 2 | 544 | |||
AK035218.1 | - | 2 | 554 | |||
AK122576.2 | - | 1 | 61 | |||
AK142298.1 | - | 2 | 412 | |||
AK147253.1 | - | 2 | 719 | |||
BC146005.1 | - | 2 | 503 | 40131308 | ||
BC146007.1 | - | 2 | 502 | 40131310 |