Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnfsf15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177371.3 | NP_796345.3 | 4 | 826 | ||
GenBank mRNA | AF520786.1 | - | 4 | 743 | ||
AY764130.1 | - | 4 | 826 | |||
BC096494.1 | - | 4 | 812 | 30740802 | ||
BC138207.1 | - | 4 | 791 | 8861226 | ||
BC138208.1 | - | 4 | 791 | 8861227 |