Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc39a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026244.2 | NP_080520.2 | 6 | 1343 | ||
GenBank mRNA | AK014732.1 | - | 6 | 1049 | ||
AK014734.1 | - | 6 | 1049 | |||
AK080462.1 | - | 6 | 1325 | |||
AK090062.1 | - | 6 | 1336 | |||
BC043112.1 | - | 6 | 1290 | |||
BC072655.1 | - | 6 | 1319 | |||
BC117754.1 | - | 6 | 937 | 40090588 | ||
BC117755.1 | - | 6 | 937 | 40090589 | ||
BC158056.1 | - | 6 | 1088 | 9088164 |