Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apol11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143686.1 | NP_001137158.1 | 4 | 551 | ||
GenBank mRNA | AK039744.1 | - | 3 | 429 | ||
AY662646.1 | - | 5 | 785 | |||
BB634068.1 | - | 3 | 428 | |||
BB653856.1 | - | 4 | 552 | |||
BC145883.1 | - | 3 | 475 | 40131186 |