Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025163.1 | NP_001020334.1 | 5 | 426 | ||
NM_001112805.1 | NP_001106276.1 | 4 | 330 | |||
NM_177888.4 | NP_808556.1 | 5 | 569 | |||
GenBank mRNA | AK039658.1 | - | 5 | 542 | ||
AK134974.1 | - | 5 | 455 | |||
AK163133.1 | - | 4 | 329 | |||
AK173164.1 | - | 4 | 350 | |||
BB603663.2 | - | 5 | 427 | |||
BC085322.1 | - | 5 | 417 | 30739758 | ||
U29511.1 | - | 2 | 118 |